missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(4-Fluorophenyl)maleamic acid, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L1039403
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-(4-Fluorophenyl)maleamic acid | |
207°C to 209°C | |
MFCD00082643 | |
2842921 | |
NRDZVHHPNZDWRA-WAYWQWQTSA-N | |
(Z)-4-(4-fluoroanilino)-4-oxobut-2-enoic acid | |
1551046 | |
96% |
60252-79-1 | |
C10H8FNO3 | |
1 g | |
z-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, 2z-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, n-4-fluorophenyl maleamic acid, 2z-3-4-fluorophenyl carbamoyl prop-2-enoic acid, 2e-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, n-4-fluorophenyl maleamidic acid, z-4-4-fluoroanilino-4-oxobut-2-enoic acid, 2z-4-4-fluoroanilino-4-oxobut-2-enoic acid, 2-butenoic acid,4-4-fluorophenyl amino-4-oxo-, 2z | |
C1=CC(=CC=C1NC(=O)C=CC(=O)O)F | |
209.176 | |
209.18 |
Chemical Identifiers
60252-79-1 | |
209.176 | |
NRDZVHHPNZDWRA-WAYWQWQTSA-N | |
1551046 | |
C1=CC(=CC=C1NC(=O)C=CC(=O)O)F |
C10H8FNO3 | |
MFCD00082643 | |
z-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, 2z-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, n-4-fluorophenyl maleamic acid, 2z-3-4-fluorophenyl carbamoyl prop-2-enoic acid, 2e-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, n-4-fluorophenyl maleamidic acid, z-4-4-fluoroanilino-4-oxobut-2-enoic acid, 2z-4-4-fluoroanilino-4-oxobut-2-enoic acid, 2-butenoic acid,4-4-fluorophenyl amino-4-oxo-, 2z | |
(Z)-4-(4-fluoroanilino)-4-oxobut-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures