missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorophenyl)benzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H55847MD

 View more versions of this product

Catalog No. AAH55847MD



Specifications

Specifications

N-(4-Fluorophenyl)benzenesulfonamide
312-63-0
MFCD00091019
AZORQGXJAXEIGC-UHFFFAOYSA-N
N-(4-fluorophenyl)benzenesulfonamide
295229
97%
250mg
C12H10FNO2S
4'-fluorobenzenesulfonanilide
C1=CC=C(C=C1)S(=O)(=O)NC2=CC=C(C=C2)F
251.275
251.28

Chemical Identifiers

312-63-0
251.275
AZORQGXJAXEIGC-UHFFFAOYSA-N
295229
C1=CC=C(C=C1)S(=O)(=O)NC2=CC=C(C=C2)F
C12H10FNO2S
MFCD00091019
4'-fluorobenzenesulfonanilide
N-(4-fluorophenyl)benzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents