missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorobenzyl)cyclopentylamine, 97%

Manufacturer:  Alfa Aesar™ H5605103

 View more versions of this product

Catalog No. AAH5605103



Description

Description

Notes
Incompatible with oxidizing agents and acids. Store at room temperature.
Specifications

Specifications

N-(4-Fluorobenzyl)cyclopentylamine
85952-73-4
MFCD03724763
cyclopentyl 4-fluorophenyl methyl amine
C1CCC(C1)NCC2=CC=C(C=C2)F
193.265
193.26
1g
C12H16FN
UN3259
AZUJIJXHVNXASI-UHFFFAOYSA-N
N-[(4-fluorophenyl)methyl]cyclopentanamine
834084
97%

Chemical Identifiers

85952-73-4
193.265
AZUJIJXHVNXASI-UHFFFAOYSA-N
834084
C1CCC(C1)NCC2=CC=C(C=C2)F
C12H16FN
MFCD03724763
cyclopentyl 4-fluorophenyl methyl amine
N-[(4-fluorophenyl)methyl]cyclopentanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents