missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorobenzyl)-4-nitrobenzamide, 97%

Chemical Identifiers

CAS 331434-68-5
Molecular Formula C14H11FN2O3
Molecular Weight (g/mol) 274.251
MDL Number MFCD00751048
InChI Key JNVXYLPECATQFV-UHFFFAOYSA-N
Synonym n-4-fluoro-benzyl-4-nitro-benzamide, n-4-fluorobenzyl-4-nitrobenzamide, n-4-fluorophenyl methyl-4-nitrobenzamide
PubChem CID 1101313
IUPAC Name N-[(4-fluorophenyl)methyl]-4-nitrobenzamide
SMILES C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)[N+](=O)[O-])F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60236MD
View Documents
Alfa Aesar™
H60236MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6023603
View Documents
Alfa Aesar™
H6023603
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

331434-68-5
274.251
JNVXYLPECATQFV-UHFFFAOYSA-N
1101313
C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)[N+](=O)[O-])F
C14H11FN2O3
MFCD00751048
n-4-fluoro-benzyl-4-nitro-benzamide, n-4-fluorobenzyl-4-nitrobenzamide, n-4-fluorophenyl methyl-4-nitrobenzamide
N-[(4-fluorophenyl)methyl]-4-nitrobenzamide

Specifications

250mg
C14H11FN2O3
n-4-fluoro-benzyl-4-nitro-benzamide, n-4-fluorobenzyl-4-nitrobenzamide, n-4-fluorophenyl methyl-4-nitrobenzamide
C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)[N+](=O)[O-])F
274.251
274.25
N-(4-Fluorobenzyl)-4-nitrobenzamide
331434-68-5
MFCD00751048
JNVXYLPECATQFV-UHFFFAOYSA-N
N-[(4-fluorophenyl)methyl]-4-nitrobenzamide
1101313
97%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures