missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorobenzyl)-4-nitrobenzamide, 97%

Manufacturer:  Alfa Aesar™ H6023603

 View more versions of this product

Catalog No. AAH6023603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-(4-Fluorobenzyl)-4-nitrobenzamide
331434-68-5
MFCD00751048
JNVXYLPECATQFV-UHFFFAOYSA-N
N-[(4-fluorophenyl)methyl]-4-nitrobenzamide
1101313
97%
1g
C14H11FN2O3
n-4-fluoro-benzyl-4-nitro-benzamide, n-4-fluorobenzyl-4-nitrobenzamide, n-4-fluorophenyl methyl-4-nitrobenzamide
C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)[N+](=O)[O-])F
274.251
274.25

Chemical Identifiers

331434-68-5
274.251
JNVXYLPECATQFV-UHFFFAOYSA-N
1101313
C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)[N+](=O)[O-])F
C14H11FN2O3
MFCD00751048
n-4-fluoro-benzyl-4-nitro-benzamide, n-4-fluorobenzyl-4-nitrobenzamide, n-4-fluorophenyl methyl-4-nitrobenzamide
N-[(4-fluorophenyl)methyl]-4-nitrobenzamide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents