missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(4-Fluorobenzyl)-4-methoxyaniline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H56102MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsAs pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment. Ethers substituted with fluorine are employed as volatile anesthetics. In metered-dose inhalers, which are used to administer some asthma medications, flurorocarbons find application as propellants.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications
N-(4-Fluorobenzyl)-4-methoxyaniline | |
C14H14FNO | |
250 mg | |
QHNUQKUZGHGTCA-UHFFFAOYSA-N | |
N-[(4-fluorophenyl)methyl]-4-methoxyaniline | |
964317 | |
97% |
356531-43-6 | |
MFCD03210757 | |
n-4-fluorobenzyl-4-methoxyaniline, n-4-fluorophenyl methyl-4-methoxyaniline, n-4-methoxyphenyl-4-fluorobenzenemethanamine | |
COC1=CC=C(C=C1)NCC2=CC=C(C=C2)F | |
231.27 | |
231.26 |
Chemical Identifiers
356531-43-6 | |
231.27 | |
QHNUQKUZGHGTCA-UHFFFAOYSA-N | |
964317 | |
COC1=CC=C(C=C1)NCC2=CC=C(C=C2)F |
C14H14FNO | |
MFCD03210757 | |
n-4-fluorobenzyl-4-methoxyaniline, n-4-fluorophenyl methyl-4-methoxyaniline, n-4-methoxyphenyl-4-fluorobenzenemethanamine | |
N-[(4-fluorophenyl)methyl]-4-methoxyaniline |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures