missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorobenzyl)-4-methoxyaniline, 97%

Manufacturer:  Alfa Aesar™ H5610203

 View more versions of this product

Catalog No. AAH5610203



Description

Description

Applications
As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment. Ethers substituted with fluorine are employed as volatile anesthetics. In metered-dose inhalers, which are used to administer some asthma medications, flurorocarbons find application as propellants.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

N-(4-Fluorobenzyl)-4-methoxyaniline
356531-43-6
MFCD03210757
QHNUQKUZGHGTCA-UHFFFAOYSA-N
N-[(4-fluorophenyl)methyl]-4-methoxyaniline
964317
97%
1g
C14H14FNO
n-4-fluorobenzyl-4-methoxyaniline
COC1=CC=C(C=C1)NCC2=CC=C(C=C2)F
231.27
231.26

Chemical Identifiers

356531-43-6
231.27
QHNUQKUZGHGTCA-UHFFFAOYSA-N
964317
COC1=CC=C(C=C1)NCC2=CC=C(C=C2)F
C14H14FNO
MFCD03210757
n-4-fluorobenzyl-4-methoxyaniline
N-[(4-fluorophenyl)methyl]-4-methoxyaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents