Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine, 98%

Chemical Identifiers

Chemical Name or Material N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine
CAS 150812-21-8
Formula Weight 261.26
MDL Number MFCD12828054
Beilstein 22723917
Synonym 2-Amino-5-[(4-fluorobenzyl)amino]-1-nitrobenzene; N-(4-Fluorobenzyl)-3-nitrobenzene-1,4-diamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6476603

  SDS
alfa aesar™
H6476603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6476606

  SDS
alfa aesar™
H6476606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 261.26
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color Red to brown
Quantity 1g
Physical Form Powder
Synonym 2-Amino-5-[(4-fluorobenzyl)amino]-1-nitrobenzene; N-(4-Fluorobenzyl)-3-nitrobenzene-1,4-diamine
Assay 98%
Chemical Name or Material N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine
CAS 150812-21-8
Beilstein 22723917
MDL Number MFCD12828054
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.