missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorobenzyl)-3-methylaniline, 97%

Manufacturer:  Alfa Aesar™ H5619403

 View more versions of this product

Catalog No. AAH5619403



Description

Description

Applications
In materials science, organofluorine compounds have been employed in many niche applications. Fluid fluoropolymers, having a low coefficient of friction, find use as specialty lubricants.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

N-(4-Fluorobenzyl)-3-methylaniline
1019518-18-3
MFCD11138269
IMKKJCAMRXSOQM-UHFFFAOYSA-N
N-[(4-fluorophenyl)methyl]-3-methylaniline
28422119
97%
1g
C14H14FN
n-4-fluorobenzyl-3-methylaniline
CC1=CC(=CC=C1)NCC2=CC=C(C=C2)F
215.271
215.27

Chemical Identifiers

1019518-18-3
215.271
IMKKJCAMRXSOQM-UHFFFAOYSA-N
28422119
CC1=CC(=CC=C1)NCC2=CC=C(C=C2)F
C14H14FN
MFCD11138269
n-4-fluorobenzyl-3-methylaniline
N-[(4-fluorophenyl)methyl]-3-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents