missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Ethylphenyl)-2-methoxybenzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 418777-17-0
Molecular Formula C16H17NO2
Molecular Weight (g/mol) 255.317
MDL Number MFCD01152864
InChI Key WLRSWQTWFMCYRS-UHFFFAOYSA-N
Synonym n-4-ethylphenyl-2-methoxybenzamide, n-4-ethylphenyl 2-methoxyphenyl carboxamide
PubChem CID 871393
IUPAC Name N-(4-ethylphenyl)-2-methoxybenzamide
SMILES CCC1=CC=C(C=C1)NC(=O)C2=CC=CC=C2OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59920MD View Documents Alfa Aesar™
H59920MD
250mg Each for $211.27
 
AAH5992003 View Documents Alfa Aesar™
H5992003
1g Each for $675.45
 
Specifications

Chemical Identifiers

418777-17-0
255.317
WLRSWQTWFMCYRS-UHFFFAOYSA-N
871393
CCC1=CC=C(C=C1)NC(=O)C2=CC=CC=C2OC
C16H17NO2
MFCD01152864
n-4-ethylphenyl-2-methoxybenzamide, n-4-ethylphenyl 2-methoxyphenyl carboxamide
N-(4-ethylphenyl)-2-methoxybenzamide

Specifications

250mg
C16H17NO2
n-4-ethylphenyl-2-methoxybenzamide, n-4-ethylphenyl 2-methoxyphenyl carboxamide
CCC1=CC=C(C=C1)NC(=O)C2=CC=CC=C2OC
255.317
274.31
N-(4-Ethylphenyl)-2-methoxybenzamide
418777-17-0
MFCD01152864
WLRSWQTWFMCYRS-UHFFFAOYSA-N
N-(4-ethylphenyl)-2-methoxybenzamide
871393
97%