missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Ethoxyphenyl)maleamic acid, 97%

$70.04 - $732.70

Chemical Identifiers

CAS 108087-84-9
Molecular Formula C12H13NO4
Formula Weight 235.24
MDL Number MFCD00020477
Synonym 4-(4-Ethoxyanilino)-4-oxobut-2-enoic acid
Chemical Name or Material N-(4-Ethoxyphenyl)maleamic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2502406 SDS Alfa Aesar™
B2502406
5g Each for $70.04
AAB2502414 SDS Alfa Aesar™
B2502414
25g Each for $250.91
AAB2502422 SDS Alfa Aesar™
B2502422
100g Each for $732.70
Specifications Description & Specifications

Chemical Identifiers

108087-84-9
235.24
4-(4-Ethoxyanilino)-4-oxobut-2-enoic acid
C12H13NO4
MFCD00020477
N-(4-Ethoxyphenyl)maleamic acid

Specifications

108087-84-9
235.24
MFCD00020477
4-(4-Ethoxyanilino)-4-oxobut-2-enoic acid
97%
C12H13NO4
188° to 189°C
5g
N-(4-Ethoxyphenyl)maleamic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning