Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Cyanophenyl)thiourea, 98%

Chemical Identifiers

Chemical Name or Material N-(4-Cyanophenyl)thiourea
CAS 3460-55-7
Molecular Formula C8H7N3S
Formula Weight 177.23
MDL Number MFCD00041175
UN Number UN3439
Beilstein 2718720
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1221903

  SDS
alfa aesar™
L1221903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1221906

  SDS
alfa aesar™
L1221906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.23
Chemical Name or Material N-(4-Cyanophenyl)thiourea
MDL Number MFCD00041175
Assay 98%
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
CAS 3460-55-7
Molecular Formula C8H7N3S
Quantity 1g
UN Number UN3439
Melting Point ≈205°C
Beilstein 2718720
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.