missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Cyanophenyl)thiourea, 98%

Manufacturer:  Alfa Aesar™ L1221906

 View more versions of this product

Catalog No. AAL1221906



Specifications

Specifications

N-(4-Cyanophenyl)thiourea
5g
C8H7N3S
UN3439
4-cyanophenyl thiourea, 1-4-cyanophenyl thiourea, 1-4-cyanophenyl-2-thiourea, n-4-cyanophenyl thiourea, 4-cyanophenyl-thiourea, acmc-1ajd4, thiourea,n-4-cyanophenyl, 4-cyanophenyl thiourea, 4-aminothioxomethyl amino benzenecarbonitrile
C1=CC(=CC=C1C#N)NC(=S)N
177.225
177.23
~205°C
3460-55-7
MFCD00041175
2718720
LFMJROANUIRGBS-UHFFFAOYSA-N
(4-cyanophenyl)thiourea
2757973
98%

Chemical Identifiers

3460-55-7
177.225
LFMJROANUIRGBS-UHFFFAOYSA-N
2757973
C1=CC(=CC=C1C#N)NC(=S)N
C8H7N3S
MFCD00041175
4-cyanophenyl thiourea, 1-4-cyanophenyl thiourea, 1-4-cyanophenyl-2-thiourea, n-4-cyanophenyl thiourea, 4-cyanophenyl-thiourea, acmc-1ajd4, thiourea,n-4-cyanophenyl, 4-cyanophenyl thiourea, 4-aminothioxomethyl amino benzenecarbonitrile
(4-cyanophenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents