missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Cyanophenyl)guanidine, 97%

$84.23 - $1,004.50

Chemical Identifiers

CAS 5637-42-3
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD16251323
InChI Key BTDGLZSKNFJBER-UHFFFAOYSA-N
Synonym 1-4-cyanophenyl guanidine
PubChem CID 17747914
IUPAC Name 2-(4-cyanophenyl)guanidine
SMILES C1=CC(=CC=C1C#N)N=C(N)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54761MD View Documents Alfa Aesar™
H54761MD
250mg Each for $84.23
 
AAH5476103 View Documents Alfa Aesar™
H5476103
1g Each for $250.73
 
AAH5476106 View Documents Alfa Aesar™
H5476106
5g Each for $1,004.50
 
Specifications

Chemical Identifiers

5637-42-3
160.18
BTDGLZSKNFJBER-UHFFFAOYSA-N
17747914
C1=CC(=CC=C1C#N)N=C(N)N
C8H8N4
MFCD16251323
1-4-cyanophenyl guanidine
2-(4-cyanophenyl)guanidine

Specifications

250mg
C8H8N4
1-4-cyanophenyl guanidine
C1=CC(=CC=C1C#N)N=C(N)N
160.18
160.18
N-(4-Cyanophenyl)guanidine
5637-42-3
MFCD16251323
BTDGLZSKNFJBER-UHFFFAOYSA-N
2-(4-cyanophenyl)guanidine
17747914
97%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures