missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Cyanophenyl)guanidine, 97%

Manufacturer:  Alfa Aesar™ H5476103

 View more versions of this product

Catalog No. AAH5476103



Specifications

Specifications

N-(4-Cyanophenyl)guanidine
5637-42-3
MFCD16251323
BTDGLZSKNFJBER-UHFFFAOYSA-N
2-(4-cyanophenyl)guanidine
17747914
97%
1g
C8H8N4
1-4-cyanophenyl guanidine
C1=CC(=CC=C1C#N)N=C(N)N
160.18
160.18

Chemical Identifiers

5637-42-3
160.18
BTDGLZSKNFJBER-UHFFFAOYSA-N
17747914
C1=CC(=CC=C1C#N)N=C(N)N
C8H8N4
MFCD16251323
1-4-cyanophenyl guanidine
2-(4-cyanophenyl)guanidine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents