missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide, TCI America™

Manufacturer:  TCI America C31885G

 View more versions of this product

Catalog No. C31885G



Specifications

Specifications

N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide
143°C
90357-53-2
MFCD03411609
HHWDZLSGDDXUSM-UHFFFAOYSA-N
N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
11149469
Crystal-Powder at 20°C
White
5g
C12H9F3N2O
3077
CC(=C)C(=O)NC1=CC(=C(C=C1)C#N)C(F)(F)F
254.212
254.21
≥98.0% (GC)

Chemical Identifiers

90357-53-2
254.212
HHWDZLSGDDXUSM-UHFFFAOYSA-N
N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
C12H9F3N2O
MFCD03411609
11149469
CC(=C)C(=O)NC1=CC(=C(C=C1)C#N)C(F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents