Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Chlorophenyl)thiourea, 98%

Chemical Identifiers

Chemical Name or Material N-(4-Chlorophenyl)thiourea
CAS 3696-23-9
Molecular Formula C7H7ClN2S
Formula Weight 186.66
MDL Number MFCD00022168
UN Number UN2811
Beilstein 973258
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1265106

  SDS
alfa aesar™
A1265106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1265114

  SDS
alfa aesar™
A1265114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.66
Molecular Formula C7H7ClN2S
RTECS Number YS7175000
Health Hazard 1 Danger
Odor Odorless
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
MDL Number MFCD00022168
Health Hazard 3 GHS P Statement
P280f-P301+P310-P302+P350-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material N-(4-Chlorophenyl)thiourea
CAS 3696-23-9
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
Assay 98%
Melting Point 177° to 182°C
UN Number UN2811
Quantity 5g
Beilstein 973258
Safety and Handling

Danger

GHS P Statement
P280f-P301+P310-P302+P350-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300
Fatal if swallowed.