missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Chlorophenyl)phthalimide, 98+%

Manufacturer:  Alfa Aesar™ A1505706

 View more versions of this product

Catalog No. AAA1505706



Specifications

Specifications

N-(4-Chlorophenyl)phthalimide
5g
C14H8ClNO2
189458
QKHKQJWODBAIMN-UHFFFAOYSA-N
2-(4-chlorophenyl)isoindole-1,3-dione
81867
≥98%
195°C to 199°C
7386-21-2
MFCD00023048
n-4-chlorophenyl phthalimide, n-p-chlorophenyl phthalimide, phthalimide, n-p-chlorophenyl, 1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl, 2-4-chlorophenyl isoindoline-1,3-dione, 2-4-chlorophenyl-1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl isoindole-1,3-dione, acmc-209osn, 5-21-10-00293 beilstein handbook reference, n-4-chlorophenyl-phthalimide
C1=CC=C2C(=C1)C(=O)N(C2=O)C3=CC=C(C=C3)Cl
257.673
257.68

Chemical Identifiers

7386-21-2
257.673
QKHKQJWODBAIMN-UHFFFAOYSA-N
81867
C1=CC=C2C(=C1)C(=O)N(C2=O)C3=CC=C(C=C3)Cl
C14H8ClNO2
MFCD00023048
n-4-chlorophenyl phthalimide, n-p-chlorophenyl phthalimide, phthalimide, n-p-chlorophenyl, 1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl, 2-4-chlorophenyl isoindoline-1,3-dione, 2-4-chlorophenyl-1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl isoindole-1,3-dione, acmc-209osn, 5-21-10-00293 beilstein handbook reference, n-4-chlorophenyl-phthalimide
2-(4-chlorophenyl)isoindole-1,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TI4267000

Documents