Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Chlorophenyl)phthalimide, 98+%

$45.09 - $507.96

Chemical Identifiers

Chemical Name or Material N-(4-Chlorophenyl)phthalimide
CAS 7386-21-2
Molecular Formula C14H8ClNO2
Formula Weight 257.68
MDL Number MFCD00023048
Beilstein 189458
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1505706

 
alfa aesar™
A1505706
5g Each for $45.09

AAA1505714

 
alfa aesar™
A1505714
25g Each for $160.08

AAA1505722

 
alfa aesar™
A1505722
100g Each for $507.96
Description & Specifications

Specifications

Formula Weight 257.68
Chemical Name or Material N-(4-Chlorophenyl)phthalimide
Assay ≥98%
Molecular Formula C14H8ClNO2
Melting Point 195° to 199°C
MDL Number MFCD00023048
CAS 7386-21-2
Beilstein 189458
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number TI4267000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.