Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Chlorophenyl)-N',N'-dimethylurea, 97%


Quantity 500g

Manufacturer: alfa aesar™  L1345536

 View more versions of this product

Catalog No. AAL1345536

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 198.65
Chemical Name or Material N-(4-Chlorophenyl)-N',N'-dimethylurea
TSCA TSCA
Assay 97%
Molecular Formula C9H11ClN2O
MDL Number MFCD00018556
CAS 150-68-5
Health Hazard 1 Warning
Melting Point 170° to 176°C
UN Number UN3077
Beilstein 2097922
Quantity 500g
Merck Index 14,6261
Synonym Monuron
Health Hazard 2 GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number YS6300000

Specifications

Chemical Name or Material N-(4-Chlorophenyl)-N',N'-dimethylurea
CAS 150-68-5
Molecular Formula C9H11ClN2O
Formula Weight 198.65
MDL Number MFCD00018556
UN Number UN3077
Beilstein 2097922
Merck Index 14,6261
Synonym Monuron
Accessories

Warning

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.