Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Chlorophenyl)-N',N'-dimethylurea, 97%

Chemical Identifiers

Chemical Name or Material N-(4-Chlorophenyl)-N',N'-dimethylurea
CAS 150-68-5
Molecular Formula C9H11ClN2O
Formula Weight 198.65
MDL Number MFCD00018556
UN Number UN3077
Beilstein 2097922
Merck Index 14,6261
Synonym Monuron
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1345514

  SDS
alfa aesar™
L1345514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1345522

  SDS
alfa aesar™
L1345522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1345536

  SDS
alfa aesar™
L1345536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.65
Chemical Name or Material N-(4-Chlorophenyl)-N',N'-dimethylurea
Molecular Formula C9H11ClN2O
MDL Number MFCD00018556
CAS 150-68-5
Health Hazard 3 GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 97%
Melting Point 170° to 176°C
UN Number UN3077
Beilstein 2097922
Merck Index 14,6261
Synonym Monuron
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number YS6300000
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.