missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Chlorophenyl)-N',N'-dimethylurea, 97%

Manufacturer:  Alfa Aesar™ L1345514

 View more versions of this product

Catalog No. AAL1345514



Specifications

Specifications

N-(4-Chlorophenyl)-N',N'-dimethylurea
25g
C9H11ClN2O
UN3077
14,6261
BMLIZLVNXIYGCK-UHFFFAOYSA-N
3-(4-chlorophenyl)-1,1-dimethylurea
8800
198.65
170°C to 176°C
150-68-5
MFCD00018556
2097922
monuron, 3-4-chlorophenyl-1,1-dimethylurea, chlorfenidim, lirobetarex, monurex, monurox, monuruon, monuuron, telvar, herbicides, monuron
CN(C)C(=O)NC1=CC=C(C=C1)Cl
198.65
CHEBI:38214
97%

Chemical Identifiers

150-68-5
198.65
BMLIZLVNXIYGCK-UHFFFAOYSA-N
8800
3-(4-chlorophenyl)-1,1-dimethylurea
C9H11ClN2O
MFCD00018556
monuron, 3-4-chlorophenyl-1,1-dimethylurea, chlorfenidim, lirobetarex, monurex, monurox, monuruon, monuuron, telvar, herbicides, monuron
CHEBI:38214
CN(C)C(=O)NC1=CC=C(C=C1)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : YS6300000

TSCA : TSCA

Documents