Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Chlorophenyl)benzenesulfonamide, 97%

Chemical Identifiers

Chemical Name or Material N-(4-Chlorophenyl)benzenesulfonamide
CAS 4750-28-1
Formula Weight 267.73
MDL Number MFCD00541864
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6003203

 
alfa aesar™
H6003203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH60032MD

 
alfa aesar™
H60032MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267.73
Chemical Name or Material N-(4-Chlorophenyl)benzenesulfonamide
Physical Form Solid
Assay 97%
MDL Number MFCD00541864
CAS 4750-28-1
Color Pale purple
Quantity 1g