missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(4-Chloro-2-butynyl)phthalimide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1631303

 View more versions of this product

Catalog No. AAL1631303



Description

Description

N-(4-Chloro-2-butynyl)phthalimide is used to make other chemicals and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(4-Chloro-2-butynyl)phthalimide is used to make other chemicals and as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
Specifications

Specifications

N-(4-Chloro-2-butynyl)phthalimide
115°C to 120°C
C12H8ClNO2
1 g
179626
Insoluble in water.
C1=CC=C2C(=C1)C(=O)N(C2=O)CC#CCCl
233.651
233.66
4819-69-6
Nearly Odorless
MFCD01318120
UN3261
n-4-chloro-2-butynyl phthalimide, 2-4-chlorobut-2-yn-1-yl isoindoline-1,3-dione, 2-4-chlorobut-2-yn-1-yl-1h-isoindole-1,3 2h-dione, 2-4-chlorobut-2-yn-1-yl isoindole-1,3-dione, acmc-20akbi, n-4-chlorobut-2-ynyl phthalimide, 2-4-chlorobut-2-ynyl isoindole-1,3-dione, 2-4-chloro-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-chloro-2-butyn-1-yl, 2-4-chlorobut-2-yn-1-yl-2,3-dihydro-1h-isoindole-1,3-dione
TXNDRPKNOXQAAO-UHFFFAOYSA-N
2-(4-chlorobut-2-ynyl)isoindole-1,3-dione
237537
97%

Chemical Identifiers

4819-69-6
233.651
TXNDRPKNOXQAAO-UHFFFAOYSA-N
237537
C1=CC=C2C(=C1)C(=O)N(C2=O)CC#CCCl
C12H8ClNO2
MFCD01318120
n-4-chloro-2-butynyl phthalimide, 2-4-chlorobut-2-yn-1-yl isoindoline-1,3-dione, 2-4-chlorobut-2-yn-1-yl-1h-isoindole-1,3 2h-dione, 2-4-chlorobut-2-yn-1-yl isoindole-1,3-dione, acmc-20akbi, n-4-chlorobut-2-ynyl phthalimide, 2-4-chlorobut-2-ynyl isoindole-1,3-dione, 2-4-chloro-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-chloro-2-butyn-1-yl, 2-4-chlorobut-2-yn-1-yl-2,3-dihydro-1h-isoindole-1,3-dione
2-(4-chlorobut-2-ynyl)isoindole-1,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only