missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Carboxyphenyl)thiourea, 97%

$70.19 - $240.12

Chemical Identifiers

CAS 7366-56-5
Molecular Formula C8H8N2O2S
Formula Weight 196.23
MDL Number MFCD00041187
Beilstein 2805627
Synonym N-Phenylthiourea-4-carboxylic acid; 4-Thioureidobenzoic acid
Chemical Name or Material N-(4-Carboxyphenyl)thiourea
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1304706 SDS Alfa Aesar™
L1304706
5g Each for $240.12
AAL1304703 SDS Alfa Aesar™
L1304703
1g Each for $70.19
Specifications Description & Specifications

Chemical Identifiers

7366-56-5
196.23
2805627
N-(4-Carboxyphenyl)thiourea
C8H8N2O2S
MFCD00041187
N-Phenylthiourea-4-carboxylic acid; 4-Thioureidobenzoic acid

Specifications

7366-56-5
196.23
MFCD00041187
5g
N-Phenylthiourea-4-carboxylic acid; 4-Thioureidobenzoic acid
97%
C8H8N2O2S
≈255°C
2805627
Odorless
N-(4-Carboxyphenyl)thiourea

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning