missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Carboxyphenyl)thiourea, 97%

Manufacturer:  Alfa Aesar™ L1304706

 View more versions of this product

Catalog No. AAL1304706



Specifications

Specifications

N-(4-Carboxyphenyl)thiourea
~255°C
7366-56-5
MFCD00041187
4-thioureidobenzoic acid, 4-carbamothioylamino benzoic acid, 4-2-thioureido benzoic acid, 4-carboxyphenyl thiourea, 1-4-carboxyphenyl-2-thiourea, 1-4-carboxypheny-2-thiourea, 4-thioureido-benzoic acid, n-4-carboxyphenyl thiourea, 4-aminothioxomethyl amino benzoic acid, 4-aminocarbonothioyl amino benzoic acid
C1=CC(=CC=C1C(=O)O)NC(=S)N
196.224
196.23
Odorless
5g
C8H8N2O2S
2805627
FOYDMXZTPAEILY-UHFFFAOYSA-N
4-(carbamothioylamino)benzoic acid
736216
97%

Chemical Identifiers

7366-56-5
196.224
FOYDMXZTPAEILY-UHFFFAOYSA-N
736216
C1=CC(=CC=C1C(=O)O)NC(=S)N
C8H8N2O2S
MFCD00041187
4-thioureidobenzoic acid, 4-carbamothioylamino benzoic acid, 4-2-thioureido benzoic acid, 4-carboxyphenyl thiourea, 1-4-carboxyphenyl-2-thiourea, 1-4-carboxypheny-2-thiourea, 4-thioureido-benzoic acid, n-4-carboxyphenyl thiourea, 4-aminothioxomethyl amino benzoic acid, 4-aminocarbonothioyl amino benzoic acid
4-(carbamothioylamino)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents