Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Carboxyphenyl)phthalimide, 97%

$58.81 - $184.72

Chemical Identifiers

Chemical Name or Material N-(4-Carboxyphenyl)phthalimide
CAS 5383-82-4
Molecular Formula C15H9NO4
Formula Weight 267.24
MDL Number MFCD00023052
Beilstein 226357
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0626803

  SDS
alfa aesar™
L0626803
1g Each for $58.81

AAL0626806

  SDS
alfa aesar™
L0626806
5g Each for $184.72
Description & Specifications

Specifications

Formula Weight 267.24
Chemical Name or Material N-(4-Carboxyphenyl)phthalimide
MDL Number MFCD00023052
Assay 97%
Molecular Formula C15H9NO4
CAS 5383-82-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 290° to 295°C
Beilstein 226357
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.