Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Bromophenyl)thiourea, 97%

Chemical Identifiers

Chemical Name or Material N-(4-Bromophenyl)thiourea
CAS 2646-30-2
Molecular Formula C7H7BrN2S
Formula Weight 231.12
MDL Number MFCD00041176
UN Number UN2811
Beilstein 2361884
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1116303

  SDS
alfa aesar™
L1116303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1116306

  SDS
alfa aesar™
L1116306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.12
Chemical Name or Material N-(4-Bromophenyl)thiourea
MDL Number MFCD00041176
CAS 2646-30-2
Health Hazard 3 GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Assay 97%
Odor Odorless
Quantity 1g
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
RTECS Number YS3500000
Melting Point 178° to 184°C
Molecular Formula C7H7BrN2S
Beilstein 2361884
Safety and Handling

Danger

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.