missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Bromophenyl)thiourea, 97%

Manufacturer:  Alfa Aesar™ L1116303

 View more versions of this product

Catalog No. AAL1116303



Specifications

Specifications

N-(4-Bromophenyl)thiourea
178°C to 184°C
2646-30-2
MFCD00041176
2361884
MRVQULNOKCOGHC-UHFFFAOYSA-N
(4-bromophenyl)thiourea
2735620
231.12
Odorless
1g
C7H7BrN2S
UN2811
1-4-bromophenyl thiourea, 4-bromophenyl thiourea, n-p-bromophenylthiourea, p-bromophenylthiourea, 1-4-bromophenyl-2-thiourea, 4-bromophenyl thiourea, thiourea, 4-bromophenyl, n-4-bromophenyl thiourea, urea, 1-p-bromophenyl-2-thio, p-bromophenyl thiourea
C1=CC(=CC=C1NC(=S)N)Br
231.111
CHEBI:34391
97%

Chemical Identifiers

2646-30-2
231.111
MRVQULNOKCOGHC-UHFFFAOYSA-N
2735620
(4-bromophenyl)thiourea
C7H7BrN2S
MFCD00041176
1-4-bromophenyl thiourea, 4-bromophenyl thiourea, n-p-bromophenylthiourea, p-bromophenylthiourea, 1-4-bromophenyl-2-thiourea, 4-bromophenyl thiourea, thiourea, 4-bromophenyl, n-4-bromophenyl thiourea, urea, 1-p-bromophenyl-2-thio, p-bromophenyl thiourea
CHEBI:34391
C1=CC(=CC=C1NC(=S)N)Br
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : YS3500000

Documents