Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Bromophenyl)maleamic acid, 97%

$67.66 - $840.77

Chemical Identifiers

Chemical Name or Material N-(4-Bromophenyl)maleamic acid
CAS 36847-86-6
Molecular Formula C10H8BrNO3
Formula Weight 270.08
MDL Number MFCD00134952
Synonym 4-(4-Bromoanilino)-4-oxobut-2-enoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2521103

  SDS
alfa aesar™
B2521103
1g Each for $67.66

AAB2521106

  SDS
alfa aesar™
B2521106
5g Each for $238.07

AAB2521114

  SDS
alfa aesar™
B2521114
25g Each for $840.77
Description & Specifications

Specifications

Formula Weight 270.08
Assay 97%
Molecular Formula C10H8BrNO3
MDL Number MFCD00134952
CAS 36847-86-6
Quantity 1g
Synonym 4-(4-Bromoanilino)-4-oxobut-2-enoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material N-(4-Bromophenyl)maleamic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.