Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Bromobutoxy)phthalimide, 98+%

$145.70 - $145.70

Chemical Identifiers

Chemical Name or Material N-(4-Bromobutoxy)phthalimide
CAS 5093-32-3
Molecular Formula C12H12BrNO3
Formula Weight 298.14
MDL Number MFCD00517028
Beilstein 1533519
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1631403

  SDS
alfa aesar™
L1631403
1g Each for $145.70
Description & Specifications

Specifications

Formula Weight 298.14
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Molecular Formula C12H12BrNO3
Beilstein 1533519
Melting Point 70° to 72°C
Chemical Name or Material N-(4-Bromobutoxy)phthalimide
MDL Number MFCD00517028
CAS 5093-32-3
Assay ≥98%
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.