Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Bromo-2-methylphenyl)maleamic acid, 97%

$1,022.86 - $1,022.86

Chemical Identifiers

Chemical Name or Material N-(4-Bromo-2-methylphenyl)maleamic acid
CAS 175205-16-0
Molecular Formula C11H10BrNO3
Formula Weight 284.11g/mol
MDL Number MFCD00027403
Synonym 4-(4-Bromo-2-methylanilino)-4-oxobut-2-enoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2520714

 
alfa aesar™
B25207-14
25g Each for $1,022.86
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material N-(4-Bromo-2-methylphenyl)maleamic acid
CAS 175205-16-0
Assay 97%
Molecular Formula C11H10BrNO3
Formula Weight 284.11g/mol
MDL Number MFCD00027403
Synonym 4-(4-Bromo-2-methylanilino)-4-oxobut-2-enoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.