Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester, 99%

$57.57 - $726.26

Chemical Identifiers

Chemical Name or Material N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester
CAS 13726-52-8
Molecular Formula C16H22N2O5
Formula Weight 322.36
MDL Number MFCD00009617
Synonym Diethyl N-(4-aminobenzoyl)-L-glutamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2230509

  SDS
alfa aesar™
B2230509
10g Each for $57.57

AAB2230518

  SDS
alfa aesar™
B2230518
50g Each for $195.88

AAB2230530

  SDS
alfa aesar™
B2230530
250g Each for $726.26
Description & Specifications

Specifications

Formula Weight 322.36
CAS 13726-52-8
Melting Point 139° to 142°C
Chemical Name or Material N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester
Quantity 10g
MDL Number MFCD00009617
Molecular Formula C16H22N2O5
Assay 99%
Optical Rotation 19° (c=1 Chloroform)
Synonym Diethyl N-(4-aminobenzoyl)-L-glutamate