Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Aminobenzoyl)-beta-alanine, 98%

$45.65 - $173.28

Chemical Identifiers

Chemical Name or Material N-(4-Aminobenzoyl)-beta-alanine
CAS 7377-08-4
Molecular Formula C10H12N2O3
Formula Weight 208.22
MDL Number MFCD00009805
Synonym 3-(4-Aminobenzamido)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2021309

  SDS
alfa aesar™
B2021309
10g Each for $45.65

AAB2021318

  SDS
alfa aesar™
B2021318
50g Each for $173.28
Description & Specifications

Specifications

Formula Weight 208.22
Quantity 10g
MDL Number MFCD00009805
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C10H12N2O3
Melting Point 150° to 154°C
CAS 7377-08-4
Synonym 3-(4-Aminobenzamido)propionic acid
Health Hazard 1 Warning
Chemical Name or Material N-(4-Aminobenzoyl)-beta-alanine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.