Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Acetylphenyl)thiourea, 98%

$268.20 - $268.20

Chemical Identifiers

Chemical Name or Material N-(4-Acetylphenyl)thiourea
CAS 71680-92-7
Molecular Formula C9H10N2OS
Formula Weight 194.26
MDL Number MFCD00041188
Beilstein 2804788
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1187206

  SDS
alfa aesar™
L1187206
5g Each for $268.20
Description & Specifications

Specifications

Formula Weight 194.26
CAS 71680-92-7
Chemical Name or Material N-(4-Acetylphenyl)thiourea
Assay 98%
MDL Number MFCD00041188
Quantity 5g
Molecular Formula C9H10N2OS
Beilstein 2804788
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Melting Point ≈215°C
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.