Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Acetylphenyl)maleimide, 98%

$75.88 - $1,139.07

Chemical Identifiers

Chemical Name or Material N-(4-Acetylphenyl)maleimide
CAS 1082-85-5
Molecular Formula C12H9NO3
Formula Weight 215.21
MDL Number MFCD00014537
Beilstein 157371
Synonym 1-(4-Acetylphenyl)-1H-pyrrole-2,5-dione; 4'-Maleimidoacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1328206

 
alfa aesar™
A1328206
5g Each for $287.85

AAA1328214

 
alfa aesar™
A1328214
25g Each for $1,139.07

AAA1328203

 
alfa aesar™
A1328203
1g Each for $75.88
Description & Specifications

Specifications

Formula Weight 215.21
Chemical Name or Material N-(4-Acetylphenyl)maleimide
Assay 98%
Molecular Formula C12H9NO3
MDL Number MFCD00014537
CAS 1082-85-5
Beilstein 157371
Synonym 1-(4-Acetylphenyl)-1H-pyrrole-2,5-dione; 4'-Maleimidoacetophenone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 154° to 158°C
Quantity 5g

Applications
N-(4-Acetylphenyl)maleimide is used as cross linking agent for reversible conjugation of thiol- containing compounds.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.