missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar H6112906

 View more versions of this product

Catalog No. AAH6112906



Specifications

Specifications

N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide
5g
C16H18FN3O3S
4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met
CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C
351.396
351.4
99%
White
147118-37-4
MFCD08458342
WOCOTUDOVSLFOB-UHFFFAOYSA-N
N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
10473133
Solid

Chemical Identifiers

147118-37-4
351.396
WOCOTUDOVSLFOB-UHFFFAOYSA-N
10473133
CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C
C16H18FN3O3S
MFCD08458342
4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met
N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

Documents