missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide, 99%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H6112914

 View more versions of this product

Catalog No. AAH6112914



Specifications

Specifications

N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide
25g
C16H18FN3O3S
4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide
CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C
351.396
351.4
99%
White
147118-37-4
MFCD08458342
WOCOTUDOVSLFOB-UHFFFAOYSA-N
N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
10473133
Solid

Chemical Identifiers

147118-37-4
351.396
WOCOTUDOVSLFOB-UHFFFAOYSA-N
10473133
CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C
C16H18FN3O3S
MFCD08458342
4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide
N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents