missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-[4-(1-Naphthyl)phenyl]-4-biphenylamine, TCI America™

$233.75 - $698.20

Chemical Identifiers

CAS 897921-59-4
Molecular Formula C28H21N
Molecular Weight (g/mol) 371.483
InChI Key DOTSEUKJBPAPGG-UHFFFAOYSA-N
Synonym N-[4-(Naphthalen-1-yl)phenyl]-[1,1′-biphenyl]-4-amine
PubChem CID 58900802
IUPAC Name N-(4-naphthalen-1-ylphenyl)-4-phenylaniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)C4=CC=CC5=CC=CC=C54
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N1053200MG View Documents TCI America
N1053200MG
200mg Each for $233.75
 
N10531G View Documents TCI America
N10531G
1g Each for $698.20
 
Specifications

Chemical Identifiers

897921-59-4
371.483
N-[4-(Naphthalen-1-yl)phenyl]-[1,1′-biphenyl]-4-amine
N-(4-naphthalen-1-ylphenyl)-4-phenylaniline
C28H21N
DOTSEUKJBPAPGG-UHFFFAOYSA-N
58900802
C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)C4=CC=CC5=CC=CC=C54

Specifications

Undesignated
200mg
C28H21N
DOTSEUKJBPAPGG-UHFFFAOYSA-N
N-(4-naphthalen-1-ylphenyl)-4-phenylaniline
58900802
Crystal-Powder at 20°C
N-[4-(1-Naphthyl)phenyl]-4-biphenylamine
100°C
897921-59-4
N-[4-(Naphthalen-1-yl)phenyl]-[1,1′-biphenyl]-4-amine
C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)C4=CC=CC5=CC=CC=C54
371.483
371.48
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No