Please login to your online account to display your discounted pricing

N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  216530050

 View more versions of this product

Catalog No. AC216530050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Quantity 5g
Color Clear colorless to light yellow
Chemical Name or Material N-[3-(Trimethoxysilyl)propyl]ethylenediamine
EINECS Number 217-164-6
Refractive Index 1.4425 to 1.4445 (20°C, 589nm)
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H22N2O3Si
Linear Formula (CH3O)3Si(CH2)3NHCH2CH2NH2
MDL Number MFCD00008173
CAS 1760-24-3
Synonym DAMO
CAS Min % 96.0
Health Hazard 1 MOISTURE SENSITIVE
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
TSCA TSCA
Formula Weight 222.36
RTECS Number KV7400000
Density 1.0300g/mL

Specifications

Chemical Name or Material N-[3-(Trimethoxysilyl)propyl]ethylenediamine
Name Note 97%
CAS 1760-24-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H22N2O3Si
Linear Formula (CH3O)3Si(CH2)3NHCH2CH2NH2
Formula Weight 222.36
MDL Number MFCD00008173
Synonym DAMO
Accessories

MOISTURE SENSITIVE

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Causes skin irritation.

SDS & Safety Documents