Learn More
N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%, Thermo Scientific Chemicals
$118.65 - $389.00
Chemical Identifiers
CAS | 1760-24-3 |
---|---|
Molecular Formula | C8H22N2O3Si |
Molecular Weight (g/mol) | 222.36 |
MDL Number | MFCD00008173 |
InChI Key | PHQOGHDTIVQXHL-UHFFFAOYSA-N |
Synonym | n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane |
PubChem CID | 15659 |
IUPAC Name | N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine |
SMILES | CO[Si](CCCNCCN)(OC)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC216531000
|
Thermo Scientific Chemicals
216531000 |
100 g | Glass bottle |
Each for $118.65
|
|
||||
AC216535000
|
Thermo Scientific Chemicals
216535000 |
500 g | Glass bottle |
Each for $389.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1760-24-3 | |
222.36 | |
PHQOGHDTIVQXHL-UHFFFAOYSA-N | |
15659 | |
CO[Si](CCCNCCN)(OC)OC |
C8H22N2O3Si | |
MFCD00008173 | |
n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane | |
N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine |
Specifications
1760-24-3 | |
100.0 | |
1.0300g/mL | |
136°C | |
96% min. (GC) | |
C8H22N2O3Si | |
(CH3O)3Si(CH2)3NHCH2CH2NH2 | |
100 g | |
n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane | |
PHQOGHDTIVQXHL-UHFFFAOYSA-N | |
N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine | |
15659 | |
97% | |
N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97% |
96.0 | |
Colorless to Yellow | |
261°C | |
Authentic | |
Glass bottle | |
1.4425 to 1.4445 | |
MFCD00008173 | |
1.03 | |
Solubility in water: hydrolysis | |
CO[Si](CCCNCCN)(OC)OC | |
222.36 | |
222.36 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 217-164-6
RTECSNumber : KV7400000
TSCA : TSCA
RUO – Research Use Only