missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-[3-(Trifluoromethyl)phenylsulfonyl]glycine, 96%, Alfa Aesar™

$39.95 - $109.03

Chemical Identifiers

CAS 85845-02-9
Molecular Formula C9H8F3NO4S
Molecular Weight (g/mol) 283.221
MDL Number MFCD03786832
InChI Key YSZHCOBVLXGMJQ-UHFFFAOYSA-N
Synonym 2-3-trifluoromethyl benzenesulfonamido acetic acid
PubChem CID 697902
IUPAC Name 2-[[3-(trifluoromethyl)phenyl]sulfonylamino]acetic acid
SMILES C1=CC(=CC(=C1)S(=O)(=O)NCC(=O)O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH34214MD View Documents Alfa Aesar
H34214MD
250mg Each for $39.95
 
AAH3421403 View Documents Alfa Aesar
H3421403
1g Each for $109.03
 
Specifications

Chemical Identifiers

85845-02-9
283.221
YSZHCOBVLXGMJQ-UHFFFAOYSA-N
697902
C1=CC(=CC(=C1)S(=O)(=O)NCC(=O)O)C(F)(F)F
C9H8F3NO4S
MFCD03786832
2-3-trifluoromethyl benzenesulfonamido acetic acid
2-[[3-(trifluoromethyl)phenyl]sulfonylamino]acetic acid

Specifications

148°C to 149°C
85845-02-9
MFCD03786832
YSZHCOBVLXGMJQ-UHFFFAOYSA-N
2-[[3-(trifluoromethyl)phenyl]sulfonylamino]acetic acid
697902
96%
250mg
C9H8F3NO4S
2-3-trifluoromethyl benzenesulfonamido acetic acid
C1=CC(=CC(=C1)S(=O)(=O)NCC(=O)O)C(F)(F)F
283.221
283.23
N-[3-(Trifluoromethyl)phenylsulfonyl]glycine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning