Please login to your online account to display your discounted pricing

Alfa Aesar™ N-[3-(Trifluoromethyl)phenyl]thiourea, 97%

Chemical Identifiers

Chemical Name or Material N-[3-(Trifluoromethyl)phenyl]thiourea
CAS 1736-70-5
Molecular Formula C8H7F3N2S
Formula Weight 220.22
MDL Number MFCD00041171
UN Number UN2811
Beilstein 2695477
Synonym N-(alpha,alpha,alpha-Trifluoro-m-tolyl)thiourea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1044403

 
alfa aesar™
L1044403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1044406

 
alfa aesar™
L1044406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 220.22
Chemical Name or Material N-[3-(Trifluoromethyl)phenyl]thiourea
Assay 97%
Molecular Formula C8H7F3N2S
MDL Number MFCD00041171
CAS 1736-70-5
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Odor Odorless
Quantity 1g
UN Number UN2811
Beilstein 2695477
Synonym N-(alpha,alpha,alpha-Trifluoro-m-tolyl)thiourea
Health Hazard 1 Danger
Melting Point 103° to 106°C
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.