missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-[3-(Trifluoromethyl)phenyl]thiourea, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar L1044406

 View more versions of this product

Catalog No. AAL1044406



Specifications

Specifications

N-[3-(Trifluoromethyl)phenyl]thiourea
103°C to 106°C
1736-70-5
MFCD00041171
2695477
WKRUQAYFMKZMPJ-UHFFFAOYSA-N
[3-(trifluoromethyl)phenyl]thiourea
735844
97%
Odorless
5g
C8H7F3N2S
UN2811
3-trifluoromethyl phenylthiourea, 1-3-trifluoromethyl phenyl thiourea, 3-trifluoromethyl phenyl thiourea, n-3-trifluoromethyl phenyl thiourea, 1-3-trifluoromethyl phenyl thiourea, thiourea, 3-trifluoromethyl phenyl, 1-3-trifluoromethyl phenyl-2-thiourea, 1-3-trifluoromethyl phenyl-2-thiourea, amino 3-trifluoromethyl phenyl amino methane-1-thione, acmc-20ah62
C1=CC(=CC(=C1)NC(=S)N)C(F)(F)F
220.213
220.22

Chemical Identifiers

1736-70-5
220.213
WKRUQAYFMKZMPJ-UHFFFAOYSA-N
735844
C1=CC(=CC(=C1)NC(=S)N)C(F)(F)F
C8H7F3N2S
MFCD00041171
3-trifluoromethyl phenylthiourea, 1-3-trifluoromethyl phenyl thiourea, 3-trifluoromethyl phenyl thiourea, n-3-trifluoromethyl phenyl thiourea, 1-3-trifluoromethyl phenyl thiourea, thiourea, 3-trifluoromethyl phenyl, 1-3-trifluoromethyl phenyl-2-thiourea, 1-3-trifluoromethyl phenyl-2-thiourea, amino 3-trifluoromethyl phenyl amino methane-1-thione, acmc-20ah62
[3-(trifluoromethyl)phenyl]thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents