Please login to your online account to display your discounted pricing

N-(3-Formyl-4-pyridinyl)-2,2-dimethylpropanamide, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00095CB

 View more versions of this product

Catalog No. MO00095CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 206.24
Quantity 250mg
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
CAS 86847-71-4
Chemical Name or Material N-(3-Formyl-4-pyridinyl)-2, 2-dimethylpropanamide
MDL Number MFCD03086208
Assay Percent Range 97%
Molecular Formula C11H14N2O2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material N-(3-Formyl-4-pyridinyl)-2, 2-dimethylpropanamide
CAS 86847-71-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H14N2O2
Formula Weight 206.24
MDL Number MFCD03086208
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.