Please login to your online account to display your discounted pricing

N-(3-Formyl-2-pyridinyl)-2,2-dimethylpropanamide, ≥90%, Maybridge

Chemical Identifiers

Chemical Name or Material N-(3-Formyl-2-pyridinyl)-2, 2-dimethylpropanamide
CAS 86847-64-5
CAS Min % 90.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C11H14N2O2
Formula Weight 206.24
MDL Number MFCD01830170
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00187DA

 
fisher scientific company
MO00187DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay 90%
Formula Weight 206.24
Quantity 1g
CAS Min % 90.0
Chemical Name or Material N-(3-Formyl-2-pyridinyl)-2, 2-dimethylpropanamide
Assay Percent Range 90%
Molecular Formula C11H14N2O2
MDL Number MFCD01830170
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber glass bottle
CAS 86847-64-5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.