Please login to your online account to display your discounted pricing

N-(3-Formyl-2-pyridinyl)-2,2-dimethylpropanamide, ≥90%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  MO00187DA

 View more versions of this product

Catalog No. MO00187DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 206.24
Assay 90%
Quantity 1g
CAS Min % 90.0
Assay Percent Range 90%
Chemical Name or Material N-(3-Formyl-2-pyridinyl)-2, 2-dimethylpropanamide
Molecular Formula C11H14N2O2
MDL Number MFCD01830170
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
CAS 86847-64-5

Specifications

Chemical Name or Material N-(3-Formyl-2-pyridinyl)-2, 2-dimethylpropanamide
CAS 86847-64-5
CAS Min % 90.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C11H14N2O2
Formula Weight 206.24
MDL Number MFCD01830170
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents