missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(3-Fluorophenyl)thiourea, 99%

Manufacturer:  Alfa Aesar™ L0953806

 View more versions of this product

Catalog No. AAL0953806



Specifications

Specifications

N-(3-Fluorophenyl)thiourea
114°C to 117°C
458-05-9
MFCD00041167
2718454
YKOBLHQALWQKTC-UHFFFAOYSA-N
(3-fluorophenyl)thiourea
2734799
99%
Odorless
5g
C7H7FN2S
UN2811
1-3-fluorophenyl-2-thiourea, 1-3-fluorophenyl thiourea, 3-fluorophenyl thiourea, n-3-fluorophenyl thiourea, amino 3-fluorophenyl amino methane-1-thione, acmc-20apbn, 1-m-fluorophenyl thiourea, 3-fluoro-phenyl-thiourea, 3-fluorophenyl thiourea, 1-3-fluorophenyl-2-thiourea
C1=CC(=CC(=C1)F)NC(=S)N
170.205
170.21

Chemical Identifiers

458-05-9
170.205
YKOBLHQALWQKTC-UHFFFAOYSA-N
2734799
C1=CC(=CC(=C1)F)NC(=S)N
C7H7FN2S
MFCD00041167
1-3-fluorophenyl-2-thiourea, 1-3-fluorophenyl thiourea, 3-fluorophenyl thiourea, n-3-fluorophenyl thiourea, amino 3-fluorophenyl amino methane-1-thione, acmc-20apbn, 1-m-fluorophenyl thiourea, 3-fluoro-phenyl-thiourea, 3-fluorophenyl thiourea, 1-3-fluorophenyl-2-thiourea
(3-fluorophenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents