Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(3-Chlorophenyl)thiourea, 98%

Chemical Identifiers

Chemical Name or Material N-(3-Chlorophenyl)thiourea
CAS 4947-89-1
Molecular Formula C7H7ClN2S
Formula Weight 186.66
MDL Number MFCD00022165
UN Number UN2811
Beilstein 2090370
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0712303

  SDS
alfa aesar™
L0712303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0712306

  SDS
alfa aesar™
L0712306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.66
Chemical Name or Material N-(3-Chlorophenyl)thiourea
Molecular Formula C7H7ClN2S
MDL Number MFCD00022165
CAS 4947-89-1
Quantity 1g
Health Hazard 1 Danger
RTECS Number YS7000000
UN Number UN2811
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
Health Hazard 3 GHS P Statement
P280f-P301+P310-P302+P350-P330-P405-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 137° to 140°C
Assay 98%
Beilstein 2090370
Safety and Handling

Danger

GHS P Statement
P280f-P301+P310-P302+P350-P330-P405-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300
Fatal if swallowed.