missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(3-Chloro-4-methylphenyl)-4-nitrobenzamide, 97%

$225.15 - $717.08

Chemical Identifiers

CAS 5344-54-7
Molecular Formula C14H11ClN2O3
Molecular Weight (g/mol) 290.703
MDL Number MFCD00171547
InChI Key WVMKIXPORUXIMJ-UHFFFAOYSA-N
Synonym n-3-chloro-4-methylphenyl-4-nitrobenzamide, 3'-chloro-4'-methyl-4-nitrobenzanilide, benzamide,n-3-chloro-4-methylphenyl-4-nitro, n-3-chloro-4-methylphenyl 4-nitrophenyl carboxamide, n-3-chloro-4-methyl-phenyl-4-nitro-benzamide
PubChem CID 295397
IUPAC Name N-(3-chloro-4-methylphenyl)-4-nitrobenzamide
SMILES CC1=C(C=C(C=C1)NC(=O)C2=CC=C(C=C2)[N+](=O)[O-])Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60232MD View Documents Alfa Aesar™
H60232MD
250mg Each for $225.15
 
AAH6023203 View Documents Alfa Aesar™
H6023203
1g Each for $717.08
 
Specifications

Chemical Identifiers

5344-54-7
290.703
WVMKIXPORUXIMJ-UHFFFAOYSA-N
295397
CC1=C(C=C(C=C1)NC(=O)C2=CC=C(C=C2)[N+](=O)[O-])Cl
C14H11ClN2O3
MFCD00171547
n-3-chloro-4-methylphenyl-4-nitrobenzamide, 3'-chloro-4'-methyl-4-nitrobenzanilide, benzamide,n-3-chloro-4-methylphenyl-4-nitro, n-3-chloro-4-methylphenyl 4-nitrophenyl carboxamide, n-3-chloro-4-methyl-phenyl-4-nitro-benzamide
N-(3-chloro-4-methylphenyl)-4-nitrobenzamide

Specifications

250mg
C14H11ClN2O3
n-3-chloro-4-methylphenyl-4-nitrobenzamide, 3'-chloro-4'-methyl-4-nitrobenzanilide, benzamide,n-3-chloro-4-methylphenyl-4-nitro, n-3-chloro-4-methylphenyl 4-nitrophenyl carboxamide, n-3-chloro-4-methyl-phenyl-4-nitro-benzamide
CC1=C(C=C(C=C1)NC(=O)C2=CC=C(C=C2)[N+](=O)[O-])Cl
290.703
290.7
N-(3-Chloro-4-methylphenyl)-4-nitrobenzamide
5344-54-7
MFCD00171547
WVMKIXPORUXIMJ-UHFFFAOYSA-N
N-(3-chloro-4-methylphenyl)-4-nitrobenzamide
295397
97%